2-(4-(Methylsulfonyl)-2-nitrophenyl)essigsäure 98%

Lieferant: Apollo Scientific

OR8252-1G
APOSOR8252-1GEA 176 EUR
APOSOR8252-1G
2-(4-(Methylsulfonyl)-2-nitrophenyl)essigsäure 98%
2-(4-(Methylsulfonyl)-2-nitrophenyl)essigsäure
Formel: C₉H₉NO₆S
Lagertemperatur: Raumtemperatur
MDL: MFCD09258635
CAS-Nummer: 929000-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR