8-Quinolinol hemisulphate salt
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
8-Hydroxyquinoline hemisulphate, 8-Quinolinone hemisulphate, Oxine sulphate, Quinolin-8-ol hemisulphate, 8-Quinolinol sulphate, 8-Hydroxyquinolinium hemisulphate, Chinosol
OR8254-10G
APOSOR8254-10GEA
29
EUR
APOSOR8254-10G
8-Quinolinol hemisulphate salt
8-Quinolinol hemisulphate salt
Formel:
C₁₈H₁₄N₂O₂·H₂SO₄ Molecular Weight: 388,39 g/mol Schmelzpunkt: 175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013095 CAS-Nummer: 134-31-6 EINECS: 205-137-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...