7-Chlorchinaldin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Chlor-2-methylchinolin
OR8258-5G
APOSOR8258-5GEA
21.9
EUR
APOSOR8258-5G
7-Chlorchinaldin
7-Chlorchinaldin
Formel:
C₁₀H₈ClN Molecular Weight: 177.63 g/mol Siedepunkt: 87 °C/0.75mmHg Schmelzpunkt: 74…78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075436 CAS-Nummer: 4965-33-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...