4-Nitro-m-kresol

Lieferant: Apollo Scientific

Synonyme: 3-Methyl-4-nitrophenol, 1-Hydroxy-3-methyl-4-nitrobenzol, 5-Hydroxy-2-nitrotoluol

OR8262-100G
APOSOR8262-100GEA 58 EUR
APOSOR8262-100G
4-Nitro-m-kresol
4-Nitro-m-kresol
Formel: C₇H₇NO₃
Molecular Weight: 153.14 g/mol
Schmelzpunkt: 127…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007333
CAS-Nummer: 2581-34-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR