cis-3-Aminocyclobutane-1-carboxylic acid, N-CBZ protected
Lieferant: Apollo Scientific
Synonyme:
cis-3-{[(Benzyloxy)carbonyl]amino}cyclobutane-1-carboxylic acid, Benzyl (cis-3-carboxycyclobut-1-yl)carbamate
OR8269-1G
APOSOR8269-1GEA
782
EUR
APOSOR8269-1G
cis-3-Aminocyclobutane-1-carboxylic acid, N-CBZ protected
cis-3-Aminocyclobutane-1-carboxylic acid, N-CBZ protected
Formel:
C₁₃H₁₅NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659967 CAS-Nummer: 1212380-76-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...