4-Hydroxy-7-methoxyquinoline-3-carboxylic acid 97%

Lieferant: Apollo Scientific

OR8293-5G
APOSOR8293-5GEA 87 EUR
APOSOR8293-5G
4-Hydroxy-7-methoxyquinoline-3-carboxylic acid 97%
4-Hydroxy-7-methoxyquinoline-3-carboxylic acid
Formel: C₁₁H₉NO₄
Molecular Weight: 219,1900 g/mol
Schmelzpunkt: 257…260 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00024004
CAS-Nummer: 28027-17-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR