4-(2-Aminoethyl)benzeneboronic acid, N-CBZ protected 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-{2-[(Benzyloxycarbonyl)amino]ethyl}benzeneboronic acid

OR8310-1G
APOSOR8310-1GEA 478 EUR
APOSOR8310-1G
4-(2-Aminoethyl)benzeneboronic acid, N-CBZ protected 98%
4-(2-Aminoethyl)benzeneboronic acid, N-CBZ protected
Formel: C₁₆H₁₈BNO₄
Molecular Weight: 299,129 g/mol
Schmelzpunkt: 161…165 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09258749
CAS-Nummer: 957034-40-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR