6-Chloro-1,3-benzothiazole-2(3H)-thione

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloro-2,3-dihydro-2-thioxo-1,3-benzothiazole

OR8320-500MG
APOSOR8320-500MGEA 21.9 EUR
APOSOR8320-500MG
6-Chloro-1,3-benzothiazole-2(3H)-thione
6-Chloro-1,3-benzothiazole-2(3H)-thione
Formel: C₇H₄ClNS₂
Lagertemperatur: Kühlschrank
MDL: MFCD02677573
CAS-Nummer: 51618-29-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR