1-(2-chloro-6-nitrophenyl)piperazine

Lieferant: Apollo Scientific

OR8329-1G
APOSOR8329-1GEA 69.6 EUR
APOSOR8329-1G
1-(2-chloro-6-nitrophenyl)piperazine
1-(2-chloro-6-nitrophenyl)piperazine
Formel: C₁₀H₁₂ClN₃O₂
Schmelzpunkt: 95…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01173865
CAS-Nummer: 332023-12-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR