1-(4-Chloro-2-nitrophenyl)piperidine-4-carboxylic acid

Lieferant: Apollo Scientific

OR8334-1G
APOSOR8334-1GEA 270 EUR
APOSOR8334-1G
1-(4-Chloro-2-nitrophenyl)piperidine-4-carboxylic acid
1-(4-Chloro-2-nitrophenyl)piperidine-4-carboxylic acid
Formel: C₁₂H₁₃ClN₂O₄
Schmelzpunkt: 123…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05664784
CAS-Nummer: 874800-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR