5-Formylfuran-2-boronsäure

Lieferant: Apollo Scientific
Gefahr

OR8338-5G
APOSOR8338-5GEA 0 EUR
APOSOR8338-5G
5-Formylfuran-2-boronsäure
5-Formylfuran-2-boronsäure
Formel: C₅H₅BO₄
Lagertemperatur: Gefrierschrank
MDL: MFCD01114696
CAS-Nummer: 27329-70-0
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR