5-(Ethylsulphonyl)-2-methoxyaniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4-(ethylsulphonyl)anisole, 5-(Ethylsulphonyl)-o-anisidine, 3-Amino-4-methoxyphenyl ethyl sulphone

OR8344-1G
APOSOR8344-1GEA 288 EUR
APOSOR8344-1G
5-(Ethylsulphonyl)-2-methoxyaniline
5-(Ethylsulphonyl)-2-methoxyaniline
Formel: C₉H₁₃NO₃S
Molecular Weight: 215,269 g/mol
Schmelzpunkt: 103…107 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00025083
CAS-Nummer: 5339-62-8
EINECS: 226-272-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR