1-[4-(Ethylsulphonyl)-2-nitrophenyl]-3-methylpiperazine hydrochloride

Lieferant: Apollo Scientific

OR8352-1G
APOSOR8352-1GEA 383 EUR
APOSOR8352-1G
1-[4-(Ethylsulphonyl)-2-nitrophenyl]-3-methylpiperazine hydrochloride
1-[4-(Ethylsulphonyl)-2-nitrophenyl]-3-methylpiperazine hydrochloride
Formel: C₁₃H₂₀ClN₃O₄S
Molecular Weight: 349,84 g/mol
Schmelzpunkt: 248…253 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD08692395
CAS-Nummer: 1172106-55-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR