1-[4-(Ethylsulphonyl)-2-nitrophenyl]-4-methylpiperazine hydrochloride

Lieferant: Apollo Scientific

OR8353-1G
APOSOR8353-1GEA 285 EUR
APOSOR8353-1G
1-[4-(Ethylsulphonyl)-2-nitrophenyl]-4-methylpiperazine hydrochloride
1-[4-(Ethylsulphonyl)-2-nitrophenyl]-4-methylpiperazine hydrochloride
Formel: C₁₃H₂₀ClN₃O₄S
Schmelzpunkt: 133…136 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD09025925
CAS-Nummer: 1170982-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR