5-Acetyl-1H-indole 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Ethanoyl-1H-indole, 1-(1H-Indol-5-yl)ethanone, 1-(1H-Indol-5-yl)ethan-1-one

OR8356-1G
APOSOR8356-1GEA 126 EUR
APOSOR8356-1G
5-Acetyl-1H-indole 97%
5-Acetyl-1H-indole
Formel: C₁₀H₉NO
Molecular Weight: 159,185 g/mol
Schmelzpunkt: 95…96 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00481875
CAS-Nummer: 53330-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR