1-[4-(Ethylsulphonyl)-2-nitrophenyl]piperidine-4-carboxamide

Lieferant: Apollo Scientific

OR8362-1G
APOSOR8362-1GEA 302 EUR
APOSOR8362-1G
1-[4-(Ethylsulphonyl)-2-nitrophenyl]piperidine-4-carboxamide
1-[4-(Ethylsulphonyl)-2-nitrophenyl]piperidine-4-carboxamide
Formel: C₁₄H₁₉N₃O₅S
Schmelzpunkt: 172…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692377
CAS-Nummer: 942474-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR