4-(3-Methyl-4-nitrophenyl)morpholin

Lieferant: Apollo Scientific

OR8376-1G
APOSOR8376-1GEA 125 EUR
APOSOR8376-1G
4-(3-Methyl-4-nitrophenyl)morpholin
4-(3-Methyl-4-nitrophenyl)morpholin
Formel: C₁₁H₁₄N₂O₃
Schmelzpunkt: 139…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692422
CAS-Nummer: 220679-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR