2-Chloro-4-(chloromethyl)thiazole

Lieferant: Apollo Scientific
Achtung

OR8403-250MG
APOSOR8403-250MGEA 50.9 EUR
APOSOR8403-250MG
2-Chloro-4-(chloromethyl)thiazole
2-Chloro-4-(chloromethyl)thiazole
Formel: C₄H₃Cl₂NS
Molecular Weight: 168,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09265510
CAS-Nummer: 5198-76-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR