(Z)-1-Ethoxyethene-2-boronic acid pinacol ester

Lieferant: Apollo Scientific
Achtung

OR8411-1G OR8411-5G
APOSOR8411-1GEA 319 EUR
APOSOR8411-1G APOSOR8411-5G
(Z)-1-Ethoxyethene-2-boronic acid pinacol ester
(Z)-1-Ethoxyethene-2-boronic acid pinacol ester
Formel: C₁₀H₁₉BO₃
Siedepunkt: 50 °C/0,3mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD09702441
CAS-Nummer: 219489-07-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR