Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
[(Tetrahydropyran-2-yloxy)methyl]phosphonic acid Diethyl ester
OR8421-5G
APOSOR8421-5GEA
401
EUR
APOSOR8421-5G
Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate
Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate
Formel:
C₁₀H₂₁O₅P Lagertemperatur: Raumtemperatur |
MDL:
MFCD09263515 CAS-Nummer: 71885-51-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...