3-Amino-4-chlorpyridin 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Chloro-3-pyridylamine, 4-Chloropyridin-3-amine

OR8424-1G
APOSOR8424-1GEA 14.7 EUR
APOSOR8424-1G
3-Amino-4-chlorpyridin 98%
3-Amino-4-chlorpyridin
Formel: C₅H₅ClN₂
Molecular Weight: 128.56 g/mol
Schmelzpunkt: 58…63 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00235124
CAS-Nummer: 20511-15-3
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR