N'-[2-(Methylsulphonyl)phenyl]hydrazine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR8430-500MG
APOSOR8430-500MGEA 306 EUR
APOSOR8430-500MG
N'-[2-(Methylsulphonyl)phenyl]hydrazine hydrochloride
N'-[2-(Methylsulphonyl)phenyl]hydrazine hydrochloride
Formel: C₇H₁₁ClN₂O₂S
Molecular Weight: 222.69 g/mol
Schmelzpunkt: 189…190 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08459596
CAS-Nummer: 912341-53-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR