1-(3-Nitropyridin-2-yl)piperidine-4-carboxamide
Lieferant: Apollo Scientific
OR8474-1G
APOSOR8474-1GEA
257
EUR
APOSOR8474-1G
1-(3-Nitropyridin-2-yl)piperidine-4-carboxamide
1-(3-Nitropyridin-2-yl)piperidine-4-carboxamide
Formel:
C₁₁H₁₄N₄O₃ Schmelzpunkt: 169…171 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00102980 CAS-Nummer: 668462-40-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...