1-(3-Nitropyridin-2-yl)piperidine-4-carboxylic acid

Lieferant: Apollo Scientific

OR8478-1G
APOSOR8478-1GEA 303 EUR
APOSOR8478-1G
1-(3-Nitropyridin-2-yl)piperidine-4-carboxylic acid
1-(3-Nitropyridin-2-yl)piperidine-4-carboxylic acid
Formel: C₁₁H₁₃N₃O₄
Schmelzpunkt: 101…103 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06208521
CAS-Nummer: 758720-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR