2-Chloro-N-(2-hydroxyethyl)-4-(1-pyrrolidinyl)benzenecarboxamide

Lieferant: Apollo Scientific

OR8498-1G
APOSOR8498-1GEA 336 EUR
APOSOR8498-1G
2-Chloro-N-(2-hydroxyethyl)-4-(1-pyrrolidinyl)benzenecarboxamide
2-Chloro-N-(2-hydroxyethyl)-4-(1-pyrrolidinyl)benzenecarboxamide
Formel: C₁₃H₁₇ClN₂O₂
Schmelzpunkt: 104…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09607906
CAS-Nummer: 952183-24-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR