2-Chloro-N-(2-hydroxyethyl)-4-(1H-1,2,4-triazol-1-yl)benzenecarboxamide

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-N-(2-hydroxyethyl)-4-(1H-1,2,4-triazol-1-yl)benzamide

OR8499-1G
APOSOR8499-1GEA 336 EUR
APOSOR8499-1G
2-Chloro-N-(2-hydroxyethyl)-4-(1H-1,2,4-triazol-1-yl)benzenecarboxamide
2-Chloro-N-(2-hydroxyethyl)-4-(1H-1,2,4-triazol-1-yl)benzenecarboxamide
Formel: C₁₁H₁₁ClN₄O₂
Molecular Weight: 266,69 g/mol
Schmelzpunkt: 140…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09607897
CAS-Nummer: 952183-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR