{1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-1H-1,2,3-triazol-4-yl}methanol

Lieferant: Apollo Scientific

Synonyme: 1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-4-(hydroxymethyl)-1H-1,2,3-triazole

OR8500-500MG
APOSOR8500-500MGEA 0 EUR
APOSOR8500-500MG
{1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-1H-1,2,3-triazol-4-yl}methanol
{1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-1H-1,2,3-triazol-4-yl}methanol
Formel: C₇H₇ClN₄OS
Molecular Weight: 230,68 g/mol
Schmelzpunkt: 109…111 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09607918
CAS-Nummer: 952183-33-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR