2-Chloro-4-(1H-1,2,4-triazol-1-yl)benzenecarbohydrazide

Lieferant: Apollo Scientific

OR8501-1G
APOSOR8501-1GEA 321 EUR
APOSOR8501-1G
2-Chloro-4-(1H-1,2,4-triazol-1-yl)benzenecarbohydrazide
2-Chloro-4-(1H-1,2,4-triazol-1-yl)benzenecarbohydrazide
Formel: C₉H₈ClN₅O
Molecular Weight: 237,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09607898
CAS-Nummer: 952183-16-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR