2-[5-Cyano-2,4-dioxo-3,4-dihydro-1(2H)-pyrimidinyl]acetic acid

Lieferant: Apollo Scientific
Achtung

OR8502-1G
APOSOR8502-1GEA 270 EUR
APOSOR8502-1G
2-[5-Cyano-2,4-dioxo-3,4-dihydro-1(2H)-pyrimidinyl]acetic acid
2-[5-Cyano-2,4-dioxo-3,4-dihydro-1(2H)-pyrimidinyl]acetic acid
Formel: C₇H₅N₃O₄
Molecular Weight: 195.14 g/mol
Schmelzpunkt: 236…239 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09607899
CAS-Nummer: 56673-29-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR