2-[5-Cyano-6-methyl-2,4-dioxo-3,4-dihydro-1(2H)-pyrimidinyl]acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Cyano-3,4-dihydro-2,4-dioxo-6-methylpyrimidin-1(2H)-yl)acetic acid

OR8503-1G
APOSOR8503-1GEA 176 EUR
APOSOR8503-1G
2-[5-Cyano-6-methyl-2,4-dioxo-3,4-dihydro-1(2H)-pyrimidinyl]acetic acid
2-[5-Cyano-6-methyl-2,4-dioxo-3,4-dihydro-1(2H)-pyrimidinyl]acetic acid
Formel: C₈H₇N₃O₄
Molecular Weight: 209,16 g/mol
Schmelzpunkt: 271…274 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09607900
CAS-Nummer: 5900-45-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR