tert-Butyl-3-(3,4-dichlorobenzyl)-3-formyltetrahydro-1(2H)-pyridinecarboxylate

Lieferant: Apollo Scientific

OR8505-500MG
APOSOR8505-500MGEA 384 EUR
APOSOR8505-500MG
tert-Butyl-3-(3,4-dichlorobenzyl)-3-formyltetrahydro-1(2H)-pyridinecarboxylate
tert-Butyl-3-(3,4-dichlorobenzyl)-3-formyltetrahydro-1(2H)-pyridinecarboxylate
Formel: C₁₈H₂₃Cl₂NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09607941
CAS-Nummer: 952183-50-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR