5-Bromo-2-(4-methylpiperazin-1-yl)pyrimidine

Lieferant: Apollo Scientific
Achtung

OR8525-500MG
APOSOR8525-500MGEA 21.9 EUR
APOSOR8525-500MG
5-Bromo-2-(4-methylpiperazin-1-yl)pyrimidine
5-Bromo-2-(4-methylpiperazin-1-yl)pyrimidine
Formel: C₉H₁₃BrN₄
Lagertemperatur: Raumtemperatur
MDL: MFCD03646018
CAS-Nummer: 141302-36-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR