(2-Phenyl-1H-imidazol-4-yl)methanol
Lieferant: Apollo Scientific
Synonyme:
4-(Hydroxymethyl)-2-phenyl-1H-imidazole
OR8526-1G
APOSOR8526-1GEA
109
EUR
APOSOR8526-1G
(2-Phenyl-1H-imidazol-4-yl)methanol
(2-Phenyl-1H-imidazol-4-yl)methanol
Formel:
C₁₀H₁₀N₂O Molecular Weight: 174.2 g/mol Schmelzpunkt: 144…146 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06202903 CAS-Nummer: 43002-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...