5-Brom-2-(methylthio)pyrimidin

Lieferant: Apollo Scientific
Gefahr

OR8529-1G OR8529-5G
APOSOR8529-1GEA 21.9 EUR
APOSOR8529-1G APOSOR8529-5G
5-Brom-2-(methylthio)pyrimidin
5-Bromo-2-(methylthio)pyrimidine
Formel: C₅H₅BrN₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00187875
CAS-Nummer: 14001-67-3
UN: 1759
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR