5-Brom-2-(methylthio)pyrimidin
Lieferant: Apollo Scientific
|
Gefahr
|
OR8529-1G
OR8529-5G
APOSOR8529-1GEA
21.9
EUR
APOSOR8529-1G
APOSOR8529-5G
5-Brom-2-(methylthio)pyrimidin
5-Bromo-2-(methylthio)pyrimidine
Formel:
C₅H₅BrN₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00187875 CAS-Nummer: 14001-67-3 UN: 1759 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...