4-(2-Bromophenyl)piperazine, N1-BOC protected
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 4-(2-bromophenyl)piperazine-1-carboxylate, 1-(2-Bromophenyl)-4-(tert-butoxycarbonyl)piperazine
OR8539-500MG
APOSOR8539-500MGEA
79.9
EUR
APOSOR8539-500MG
4-(2-Bromophenyl)piperazine, N1-BOC protected
4-(2-Bromophenyl)piperazine, N1-BOC protected
Formel:
C₁₅H₂₁BrN₂O₂ Molecular Weight: 341,243 g/mol Dichte: 1,332 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD08064040 CAS-Nummer: 494773-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...