5-Bromo-4-methoxy-2-(piperazin-1-yl)pyrimidine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-4-methoxy-2-(piperazin-1-yl)-1,3-diazine, 1-(5-Bromo-4-methoxypyrimidin-2-yl)piperazine

OR8543-500MG
APOSOR8543-500MGEA 190 EUR
APOSOR8543-500MG
5-Bromo-4-methoxy-2-(piperazin-1-yl)pyrimidine
5-Bromo-4-methoxy-2-(piperazin-1-yl)pyrimidine
Formel: C₉H₁₃BrN₄O
Molecular Weight: 273,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781226
CAS-Nummer: 885267-38-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR