4-Bromthiazol 97%

Lieferant: Apollo Scientific
Gefahr

OR8552-25G
APOSOR8552-25GEA 115 EUR
APOSOR8552-25G
4-Bromthiazol 97%
4-Bromothiazole
Formel: C₃H₂BrNS
Lagertemperatur: Kühlschrank
MDL: MFCD06657592
CAS-Nummer: 34259-99-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR