7-Methoxyisochinolin 96%
Lieferant: Apollo Scientific
|
Achtung
|
OR8563-1G
APOSOR8563-1GEA
69.6
EUR
APOSOR8563-1G
7-Methoxyisochinolin 96%
7-Methoxyisoquinoline
Formel:
C₁₀H₉NO Schmelzpunkt: 48…50 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09030747 CAS-Nummer: 39989-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...