8-Methoxyisochinolin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR8564-1G
OR8564-250MG
OR8564-5G
APOSOR8564-1GEA
326
EUR
APOSOR8564-1G
APOSOR8564-250MG
APOSOR8564-5G
8-Methoxyisochinolin 98%
8-Methoxyisoquinoline
Formel:
C₁₀H₉NO Schmelzpunkt: 48…52 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258766 CAS-Nummer: 1723-70-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...