3-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester 97%
Lieferant: Apollo Scientific
Synonyme:
3-[Methoxy(methyl)carbamoyl]benzeneboronic acid, pinacol ester
OR8567-1G
APOSOR8567-1GEA
176
EUR
APOSOR8567-1G
3-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester 97%
3-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester
Formel:
C₁₅H₂₂BNO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258775 CAS-Nummer: 957061-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...