8-Bromisochinolin 95%

Lieferant: Apollo Scientific
Achtung

OR8569-1G
APOSOR8569-1GEA 21.9 EUR
APOSOR8569-1G
8-Bromisochinolin 95%
8-Bromoisoquinoline
Formel: C₉H₆BrN
Molecular Weight: 208,0600 g/mol
Schmelzpunkt: 73…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04973298
CAS-Nummer: 63927-22-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR