3-(Methoxycarbonyl)-5-nitrobenzeneboronic acid, pinacol ester 98%

Lieferant: Apollo Scientific

OR8574-1G
APOSOR8574-1GEA 36.4 EUR
APOSOR8574-1G
3-(Methoxycarbonyl)-5-nitrobenzeneboronic acid, pinacol ester 98%
3-(Methoxycarbonyl)-5-nitrobenzeneboronic acid, pinacol ester
Formel: C₁₄H₁₈BNO₆
Lagertemperatur: Raumtemperatur
MDL: MFCD09258774
CAS-Nummer: 957061-12-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR