2-Methoxy-5-methyl-3-nitropyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR8579-1G
APOSOR8579-1GEA
69.6
EUR
APOSOR8579-1G
2-Methoxy-5-methyl-3-nitropyridin 98%
2-Methoxy-5-methyl-3-nitropyridine
Formel:
C₇H₈N₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258778 CAS-Nummer: 33252-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...