2-Brom-5-(4-morpholino)pyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(5-Bromopyridin-2-yl)morpholine, 4-(5-Bromo-2-pyridyl)morpholine, 4-(5-Bromo-pyridin-2-yl)-morpholine

OR8597-1G
APOSOR8597-1GEA 21.9 EUR
APOSOR8597-1G
2-Brom-5-(4-morpholino)pyridin
2-Brom-5-(4-morpholino)pyridin
Formel: C₉H₁₁BrN₂O
Molecular Weight: 243,1004 g/mol
Schmelzpunkt: 81…83 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02681930
CAS-Nummer: 200064-11-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR