5-Bromo-2-(pyrrolidin-1-yl)pyrimidine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(5-Bromopyrimidin-2-yl)pyrrolidine, 5-Bromo-2-(pyrrolidin-1-yl)-1,3-diazine
OR8601-1G
APOSOR8601-1GEA
21.9
EUR
APOSOR8601-1G
5-Bromo-2-(pyrrolidin-1-yl)pyrimidine
5-Bromo-2-(pyrrolidin-1-yl)pyrimidine
Formel:
C₈H₁₀BrN₃ Molecular Weight: 228,089 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00483243 CAS-Nummer: 446286-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...