4-Bromothiophene-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR8605-1G
APOSOR8605-1GEA
0
EUR
APOSOR8605-1G
4-Bromothiophene-3-carboxylic acid
4-Bromothiophene-3-carboxylic acid
Formel:
C₅H₃BrO₂S Schmelzpunkt: 158…163 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01233020 CAS-Nummer: 16694-17-0 EINECS: 240-739-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...