Ethyl-4-acetyl-5-methylisoxazole-3-carboxylate

Lieferant: Apollo Scientific
Achtung

OR8623-1G
APOSOR8623-1GEA 77 EUR
APOSOR8623-1G
Ethyl-4-acetyl-5-methylisoxazole-3-carboxylate
Ethyl-4-acetyl-5-methylisoxazole-3-carboxylate
Formel: C₉H₁₁NO₄
Molecular Weight: 197.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173953
CAS-Nummer: 15911-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR