2-Amino-5-chlor-3-nitropyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chlor-3-nitropyridin-2-amin, 5-Chlor-3-nitro-2-pyridylamin

OR8629-25G OR8629-100G
APOSOR8629-25GEA 59.9 EUR
APOSOR8629-25G APOSOR8629-100G
2-Amino-5-chlor-3-nitropyridin
2-Amino-5-chlor-3-nitropyridin
Formel: C₅H₄ClN₃O₂
Molecular Weight: 173.56 g/mol
Schmelzpunkt: 193…197 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00092011
CAS-Nummer: 5409-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR