7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carbonyl chloride 90%

Lieferant: Apollo Scientific

OR8642-1G
APOSOR8642-1GEA 248 EUR
APOSOR8642-1G
7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carbonyl chloride 90%
7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carbonyl chloride
Formel: C₇H₄BrClO₃S
Molecular Weight: 283,527 g/mol
Schmelzpunkt: 74,5…76.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08690305
CAS-Nummer: 886851-53-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR